Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113684.1 | NP_001107156.1 | 12 - 13 | 1518 | 106 | |
XM_012967939.2 | XP_012823393.1 | 13 - 14 | 1560 | 106 | ||
GenBank mRNA | BC155689.1 | - | 12 - 13 | 1518 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |