Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113684.1 | NP_001107156.1 | 29 - 30 | 3821 | 96 | |
XM_012967939.2 | XP_012823393.1 | 30 - 31 | 3863 | 96 | ||
GenBank mRNA | BC155689.1 | - | 29 - 30 | 3821 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |