Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113684.1 | NP_001107156.1 | 37 - 38 | 4646 | 76 | |
XM_012967939.2 | XP_012823393.1 | 38 - 39 | 4688 | 76 | ||
GenBank mRNA | BC155689.1 | - | 37 - 38 | 4646 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |