Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr1h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113687.1 | NP_001107159.1 | 7 - 8 | 1092 | 103 | |
XM_012952859.1 | XP_012808313.1 | 8 - 9 | 1387 | 103 | ||
XM_012952882.1 | XP_012808336.1 | 8 - 9 | 1394 | 103 | ||
GenBank mRNA | BC157257.1 | - | 7 - 8 | 1092 | 103 | |
BC170929.1 | - | 7 - 8 | 1092 | 103 | ||
BC170931.1 | - | 7 - 8 | 1092 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |