Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113691.1 | NP_001107163.1 | 3 - 4 | 226 | 75 | |
GenBank mRNA | AM041995.1 | - | 3 - 4 | 226 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Xenopus tropicalis Probe spans exons |