Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113835.1 | NP_001107307.1 | 12 - 13 | 1670 | 71 | |
XM_012964243.2 | XP_012819697.1 | 10 - 11 | 2580 | 71 | ||
GenBank mRNA | BC155702.1 | - | 12 - 13 | 1670 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |