Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113835.1 | NP_001107307.1 | 5 - 6 | 582 | 70 | |
XM_012964243.2 | XP_012819697.1 | 3 - 4 | 1482 | 70 | ||
GenBank mRNA | BC155702.1 | - | 5 - 6 | 582 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |