Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113835.1 | NP_001107307.1 | 7 - 8 | 1016 | 73 | |
XM_012964243.2 | XP_012819697.1 | 5 - 6 | 1917 | 74 | ||
GenBank mRNA | BC155702.1 | - | 7 - 8 | 1016 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |