Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113856.1 | NP_001107328.1 | 3 - 4 | 188 | 93 | |
XM_012962679.2 | XP_012818133.1 | 3 - 4 | 246 | 93 | ||
XM_012962680.2 | XP_012818134.1 | 3 - 4 | 324 | 93 | ||
XM_012962681.2 | XP_012818135.1 | 3 - 4 | 324 | 93 | ||
GenBank mRNA | BC077028.1 | - | 3 - 4 | 185 | 93 | |
BC156016.1 | - | 3 - 4 | 188 | 93 | ||
BC171191.1 | - | 3 - 4 | 186 | 93 | ||
BC171193.1 | - | 3 - 4 | 186 | 93 | ||
CT025297.2 | - | 2 - 3 | 162 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |