Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113868.1 | NP_001107340.1 | 6 - 7 | 511 | 70 | |
GenBank mRNA | BC157148.1 | - | 6 - 7 | 511 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |