Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113883.1 | NP_001107355.1 | 1 - 2 | 786 | 92 | |
GenBank mRNA | BC157202.1 | - | 1 - 2 | 786 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Xenopus tropicalis Probe spans exons |