Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kiaa0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113902.1 | NP_001107374.1 | 488 | 70 | ||
GenBank mRNA | BC157243.1 | - | 488 | 70 | ||
BC171214.1 | - | 477 | 70 | |||
BC171216.1 | - | 477 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |