Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kiaa0930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113902.1 | NP_001107374.1 | 1007 | 80 | ||
GenBank mRNA | BC157243.1 | - | 1007 | 80 | ||
BC171214.1 | - | 996 | 80 | |||
BC171216.1 | - | 996 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |