Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113906.1 | NP_001107378.1 | 3 - 4 | 982 | 86 | |
GenBank mRNA | BC157250.1 | - | 3 - 4 | 982 | 86 | |
BC170962.1 | - | 3 - 4 | 679 | 86 | ||
BC170964.1 | - | 3 - 4 | 679 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |