Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: echdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113907.1 | NP_001107379.1 | 1 - 2 | 281 | 64 | |
GenBank mRNA | BC157262.1 | - | 1 - 2 | 281 | 64 | |
BC171144.1 | - | 1 - 2 | 214 | 64 | ||
BC171148.1 | - | 1 - 2 | 214 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |