Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113911.1 | NP_001107383.1 | 4 - 5 | 430 | 85 | |
GenBank mRNA | BC157275.1 | - | 4 - 5 | 430 | 85 | |
BC170969.1 | - | 4 - 5 | 405 | 85 | ||
BC171299.1 | - | 4 - 5 | 405 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |