Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113911.1 | NP_001107383.1 | 5 - 6 | 502 | 101 | |
GenBank mRNA | BC157275.1 | - | 5 - 6 | 502 | 101 | |
BC170969.1 | - | 5 - 6 | 477 | 101 | ||
BC171299.1 | - | 5 - 6 | 477 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |