Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113951.1 | NP_001107423.1 | 6 - 7 | 770 | 72 | |
GenBank mRNA | BC157553.1 | - | 6 - 7 | 770 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |