Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114031.1 | NP_001107503.1 | 7 - 8 | 1514 | 94 | |
XM_012964257.2 | XP_012819711.1 | 94 | ||||
GenBank mRNA | BC157661.1 | - | 7 - 8 | 1514 | 94 | |
CR926350.2 | - | 1473 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |