Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114041.1 | NP_001107513.1 | 1 - 2 | 270 | 101 | |
GenBank mRNA | BC157676.1 | - | 1 - 2 | 270 | 101 | |
BC171088.1 | - | 1 - 2 | 215 | 101 | ||
BC171092.1 | - | 1 - 2 | 215 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |