Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114041.1 | NP_001107513.1 | 2 - 3 | 414 | 76 | |
GenBank mRNA | BC157676.1 | - | 2 - 3 | 414 | 76 | |
BC171088.1 | - | 2 - 3 | 359 | 76 | ||
BC171092.1 | - | 2 - 3 | 359 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |