Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114070.1 | NP_001107542.1 | 2 - 3 | 190 | 87 | |
GenBank mRNA | BC157800.1 | - | 2 - 3 | 190 | 87 | |
BC171026.1 | - | 2 - 3 | 181 | 87 | ||
BC171080.1 | - | 2 - 3 | 181 | 87 | ||
CU075862.1 | - | 2 - 3 | 151 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |