Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114203.1 | NP_001107675.1 | 921 | 69 | ||
GenBank mRNA | BC087802.1 | - | 897 | 69 | ||
BC158507.1 | - | 921 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |