Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114206.1 | NP_001107678.1 | 5 - 6 | 486 | 84 | |
GenBank mRNA | BC088048.1 | - | 5 - 6 | 454 | 84 | |
BC135180.1 | - | 5 - 6 | 460 | 84 | ||
BC153700.1 | - | 5 - 6 | 486 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |