Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114234.1 | NP_001107706.1 | 2 - 3 | 269 | 96 | |
GenBank mRNA | BC158276.1 | - | 2 - 3 | 269 | 96 | |
CR942691.2 | - | 2 - 3 | 262 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |