Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apoo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114243.1 | NP_001107715.1 | 5 - 6 | 391 | 94 | |
XM_012956316.1 | XP_012811770.1 | 5 - 6 | 408 | 94 | ||
XM_012956317.2 | XP_012811771.1 | 5 - 6 | 446 | 94 | ||
GenBank mRNA | BC135301.1 | - | 4 - 5 | 376 | 94 | |
BC155970.1 | - | 5 - 6 | 391 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |