Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114245.1 | NP_001107717.1 | 5 - 6 | 383 | 101 | |
GenBank mRNA | BC158545.1 | - | 5 - 6 | 383 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |