Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114246.1 | NP_001107718.1 | 525 | 66 | ||
GenBank mRNA | BC158525.1 | - | 525 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |