Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114248.1 | NP_001107720.1 | 4 - 5 | 443 | 77 | |
GenBank mRNA | BC158407.1 | - | 4 - 5 | 443 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |