Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cntfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114251.1 | NP_001107723.1 | 804 | 69 | ||
GenBank mRNA | BC158286.1 | - | 804 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis |