Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114489.1 | NP_001107961.1 | 3 - 4 | 537 | 77 | |
XM_012971718.2 | XP_012827172.1 | 4 - 5 | 660 | 77 | ||
XM_018097721.1 | XP_017953210.1 | 2 - 3 | 494 | 77 | ||
GenBank mRNA | BC088607.1 | - | 3 - 4 | 374 | 77 | |
BC118711.1 | - | 3 - 4 | 514 | 77 | ||
BC159128.1 | - | 3 - 4 | 537 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |