Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emsy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004912217.3 | XP_004912274.1 | 3 - 4 | 665 | 90 | |
XM_004912218.3 | XP_004912275.1 | 3 - 4 | 664 | 90 | ||
XM_004912219.3 | XP_004912276.1 | 3 - 4 | 665 | 90 | ||
XM_004912220.3 | XP_004912277.1 | 3 - 4 | 664 | 90 | ||
GenBank mRNA | BC135733.1 | - | 3 - 4 | 279 | 90 | |
CT027941.2 | - | 2 - 3 | 203 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |