Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122807.1 | NP_001116279.1 | 5 - 6 | 632 | 81 | |
GenBank mRNA | BC161222.1 | - | 5 - 6 | 632 | 81 | |
CU075789.1 | - | 5 - 6 | 625 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |