Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122812.1 | NP_001116284.1 | 2 - 3 | 475 | 82 | |
GenBank mRNA | BC157209.1 | - | 2 - 3 | 475 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |