Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123408.1 | NP_001116880.1 | 1 - 2 | 706 | 100 | |
GenBank mRNA | BC161171.1 | - | 1 - 2 | 706 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |