Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zranb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123428.1 | NP_001116900.1 | 5 - 6 | 1385 | 87 | |
XM_018095246.1 | XP_017950735.1 | 6 - 7 | 1540 | 87 | ||
XM_018095247.1 | XP_017950736.1 | 5 - 6 | 1277 | 87 | ||
XM_018095248.1 | XP_017950737.1 | 5 - 6 | 1328 | 87 | ||
GenBank mRNA | BC161086.1 | - | 5 - 6 | 1385 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |