Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123431.2 | NP_001116903.2 | 3 - 4 | 580 | 100 | |
GenBank mRNA | BC161571.1 | - | 3 - 4 | 454 | 100 | |
CX822877.1 | - | 3 - 4 | 580 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |