Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123431.2 | NP_001116903.2 | 4 - 5 | 779 | 95 | |
GenBank mRNA | BC161571.1 | - | 4 - 5 | 653 | 95 | |
CX822877.1 | - | 4 - 5 | 779 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |