Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgals2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123441.1 | NP_001116913.1 | 2 - 3 | 114 | 63 | |
GenBank mRNA | BC161334.1 | - | 2 - 3 | 114 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |