Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: best4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123442.1 | NP_001116914.1 | 7 - 8 | 1122 | 83 | |
GenBank mRNA | BC161756.1 | - | 7 - 8 | 1122 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |