Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ikzf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123458.1 | NP_001116930.1 | 6 - 7 | 929 | 57 | |
XM_012970035.2 | XP_012825489.1 | 6 - 7 | 1100 | 57 | ||
XM_012970036.2 | XP_012825490.1 | 6 - 7 | 1037 | 57 | ||
XM_012970038.2 | XP_012825492.1 | 6 - 7 | 1026 | 57 | ||
XM_012970039.2 | XP_012825493.1 | 6 - 7 | 1089 | 57 | ||
XM_012970040.2 | XP_012825494.1 | 6 - 7 | 958 | 57 | ||
XM_012970042.2 | XP_012825496.1 | 6 - 7 | 942 | 57 | ||
XM_012970043.2 | XP_012825497.1 | 5 - 6 | 908 | 57 | ||
XM_012970044.2 | XP_012825498.1 | 5 - 6 | 843 | 57 | ||
XM_012970045.2 | XP_012825499.1 | 5 - 6 | 827 | 57 | ||
XM_012970046.1 | XP_012825500.1 | 4 - 5 | 594 | 57 | ||
XM_018097034.1 | XP_017952523.1 | 6 - 7 | 828 | 57 | ||
XM_018097035.1 | XP_017952524.1 | 6 - 7 | 1037 | 57 | ||
XM_018097036.1 | XP_017952525.1 | 6 - 7 | 1036 | 57 | ||
XM_018097037.1 | XP_017952526.1 | 6 - 7 | 1037 | 57 | ||
XM_018097038.1 | XP_017952527.1 | 6 - 7 | 958 | 57 | ||
GenBank mRNA | BC161078.1 | - | 6 - 7 | 929 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |