Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123491.1 | NP_001116963.1 | 6 - 7 | 1982 | 77 | |
XM_012962698.2 | XP_012818152.1 | 7 - 8 | 2042 | 77 | ||
XM_012962699.2 | XP_012818153.1 | 6 - 7 | 2069 | 77 | ||
XM_012962700.2 | XP_012818154.1 | 7 - 8 | 2042 | 77 | ||
XM_012962701.2 | XP_012818155.1 | 7 - 8 | 2042 | 77 | ||
XM_012962702.2 | XP_012818156.1 | 7 - 8 | 2042 | 77 | ||
XM_012962703.2 | XP_012818157.1 | 7 - 8 | 2042 | 77 | ||
XM_012962704.2 | XP_012818158.1 | 7 - 8 | 2042 | 77 | ||
XM_018093774.1 | XP_017949263.1 | 7 - 8 | 2036 | 77 | ||
GenBank mRNA | BC161609.1 | - | 6 - 7 | 1982 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |