Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ammecr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123494.1 | NP_001116966.1 | 3 - 4 | 748 | 96 | |
GenBank mRNA | BC161496.1 | - | 3 - 4 | 748 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |