Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126064.1 | NP_001119536.1 | 6 - 7 | 2518 | 121 | |
GenBank mRNA | BC166334.1 | - | 6 - 7 | 2518 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |