Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126069.1 | NP_001119541.1 | 6 - 7 | 1420 | 102 | |
XM_012968579.2 | XP_012824033.1 | 6 - 7 | 1734 | 102 | ||
XM_012968580.1 | XP_012824034.1 | 6 - 7 | 1463 | 102 | ||
XM_012968581.2 | XP_012824035.1 | 5 - 6 | 1389 | 102 | ||
XM_018096299.1 | XP_017951788.1 | 6 - 7 | 1487 | 102 | ||
GenBank mRNA | BC121298.1 | - | 6 - 7 | 1407 | 102 | |
BC166063.1 | - | 6 - 7 | 1420 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |