Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126077.1 | NP_001119549.1 | 4 - 5 | 543 | 74 | |
XM_018091055.1 | XP_017946544.1 | 4 - 5 | 625 | 74 | ||
XM_018091056.1 | XP_017946545.1 | 4 - 5 | 510 | 74 | ||
GenBank mRNA | BC166190.1 | - | 4 - 5 | 543 | 74 | |
CT027907.2 | - | 4 - 5 | 481 | 74 | ||
CU024923.1 | - | 4 - 5 | 479 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |