Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: timm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126566.1 | NP_001120038.1 | 2 - 3 | 307 | 61 | |
GenBank mRNA | BC158346.1 | - | 2 - 3 | 307 | 61 | |
CT030353.1 | - | 2 - 3 | 348 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |