Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126608.1 | NP_001120080.1 | 1 - 2 | 149 | 101 | |
GenBank mRNA | BC158519.1 | - | 1 - 2 | 149 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |