Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126608.1 | NP_001120080.1 | 8 - 9 | 961 | 64 | |
GenBank mRNA | BC158519.1 | - | 8 - 9 | 961 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |