Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126655.1 | NP_001120127.1 | 6 - 7 | 757 | 96 | |
GenBank mRNA | BC159037.1 | - | 6 - 7 | 757 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |